Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 26° Baume’ (Aqua Ammonia), Reagents

Catalog No. p-200021152
Click to view available options
Quantity:
500 mL
2.5 L
4 L
15 gal.

Ammonium Hydroxide, 26° Baume’ (Aqua Ammonia), Technical, for general laboratory use. Store below 30°C. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5, 1336-21-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Aqua Ammonia
IUPAC Name water
SMILES O

Specifications

pH 13.6
Color Colorless
Boiling Point 36°C
Quantity 15 gal.
Packaging Poly Drum
Specific Gravity 0.9
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide, 26° Baume’ (Aqua Ammonia)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.