Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 28.0 to 30.0%, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
Glass Bottle
Glass Bottle, Single ship
Poly Bottle
PVC Coated Glass Bottle
1336-21-6
H5NO
35.046
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh
14923
CHEBI:18219
azanium;hydroxide
[NH4+].[OH-]
1336-21-6
H5NO
35.046
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh
14923
CHEBI:18219
azanium;hydroxide
[NH4+].[OH-]

High quality chemicals for laboratory and specialized industrial use

Quantity 500mL
Formula Weight 35.05
Percent Purity 28.0 to 30.0%
Grade BAKER ANALYZED™ ACS Reagent
Packaging Glass Bottle
Specific Gravity 1L = 0.90kg
Chemical Name or Material Ammonium Hydroxide