Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 28.0 to 30.0%, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
Glass Bottle
Glass Bottle, Single ship
Poly Bottle
PVC Coated Glass Bottle

Chemical Identifiers

1336-21-6
H5NO
35.046
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
CHEBI:18219
azanium;hydroxide
[NH4+].[OH-]

High quality chemicals for laboratory and specialized industrial use

Quantity 500mL
Formula Weight 35.05
Packaging Glass Bottle, Single ship
Specific Gravity 1L = 0.90kg
Chemical Name or Material Ammonium Hydroxide
Assay 28.0 to 30.0%
Grade Baker Analyzed™ ACS Reagent