Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 5N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Cubetainer
Narrow Mouth Polyethylene Bottle
1336-21-6
H5NO
35.046
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh
14923
CHEBI:18219
azanium;hydroxide
[NH4+].[OH-]
1336-21-6
H5NO
35.046
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh
14923
CHEBI:18219
azanium;hydroxide
[NH4+].[OH-]

High quality chemicals for laboratory and specialized industrial use

Quantity 1L
Formula Weight 35.05
Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Polyethylene Bottle
Specific Gravity 1L = 0.90kg
Chemical Name or Material Ammonium Hydroxide, 5N Volumetric Solution