Ammonium Hydroxide, ACS, 28.0-30.0%NH™, Alfa Aesar

Manufacturer: Alfa Aesar 33285D9

Quantity 5 x 1kg

 View more versions of this product

Catalog No. AA33285D9

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1336-21-6
    Physical Form Liquid
    Assay Percent Range 28.0-30.0% NH3
    Molecular Formula NH4OH
    Formula Weight 35.05
    MDL Number MFCD00066650
    Chemical Name or Material Ammonium hydroxide
    Grade ACS Grade

    Specifications

    Formula Weight 35.05
    Quantity 5 x 1kg
    Color Colorless
    DOT Information DOT Class: 8, Packaging Group: III
    EINECS Number 215-647-6
    Physical Form Liquid
    Assay Percent Range 28.0-30.0% NH3
    MDL Number MFCD00066650
    CAS 1336-21-6
    Health Hazard 1 H314-H318-H400-H302
    Appearance Colorless liquid
    Molecular Formula NH4OH
    Odor Ammonia-like
    Chemical Name or Material Ammonium hydroxide
    Grade ACS Grade

    H314-H318-H400-H302

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit