Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Hydroxide ACS, MilliporeSigma™
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Poly Coated Glass Bottle
Specifications
Assay Percent Range | 28 to 30% |
Linear Formula | NH4OH [NH3 in water] |
Density | 0.90g/cm3 (20°C) |
Quantity | 500 mL |
Explosion Limit | 15.4 to 33.6%(V) |
Formula Weight | 35.05 |
Grade | ACS/EP/Reagent |
Packaging | Glass Bottle |
Vapor Pressure | 635 hPa (20°C) |
Chemical Name or Material | Ammonium Hydroxide, EMSURE™, For Analysis |
Safety and Handling
Recommended Storage | Store below +25°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction