Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium hydroxide, ACS reagent, 28-30% solution in water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
25mL
500mL
2.5L
10L
1336-21-6, 7732-18-5
H5NO
35.046
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
14923
CHEBI:18219
azanium;hydroxide
[NH4+].[OH-]
1336-21-6, 7732-18-5
H5NO
35.046
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
14923
CHEBI:18219
azanium;hydroxide
[NH4+].[OH-]

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Additional Information Vapor Pressure: 500hPa at 20°C
Carbon Dioxide 0.002% max.
Chlorine (Cl) 0.5ppm max.
Linear Formula NH4OH
Density 0.9100g/mL
Fieser 05,15
Merck Index 15, 491
Solubility Information Solubility in water: soluble.
Heavy Metals (as Pb) 0.5ppm max.
Iron (Fe) 0.2ppm max.
Formula Weight 35.05
Physical Form Free From Suspended Matter or Sediment
Percent Purity 28.0 to 30.0% (HCl-titration (as NH3))
Grade ACS Reagent
Packaging Glass Bottle
Phosphate 2ppm max.
Nitrate 2ppm max.
Residue after Ignition 0.002% max.
Specific Gravity 0.91
Substances Reducing Permanganate Passes Test
Sulfate 2ppm max.
Melting Point -57.0°C
Boiling Point 38.0°C
Color Colorless
Quantity 2.5L
Chemical Name or Material Ammonium hydroxide
Show More Show Less

GHS P Statement
May cause respiratory irritation.
Very toxic to aquatic life.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 215-647-6

RTECSNumber : BQ9625000

TSCA : TSCA