Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium hydroxide, for analysis, 25% free ammonia, solution in water, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
1336-21-6,7732-18-5
H5NO
35.046
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh
14923
CHEBI:18219
azanium;hydroxide
[NH4+].[OH-]
1336-21-6,7732-18-5
H5NO
35.046
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh
14923
CHEBI:18219
azanium;hydroxide
[NH4+].[OH-]
Quantity 1L
Appearance of Solution Clear and colorless
Barium (Ba) 0.0001% max.
Bismuth (Bi) 0.0001% max.
Cadmium (Cd) 0.000005% max.
Calcium (Ca) 0.0001% max.
Carbonate 0.001% max.
Chloride 0.00005% max.
Chromium (Cr) 0.000005% max.
Concentration 25.0% min. free ammonia
Cobalt (Co) 0.00001% max.
Linear Formula NH4OH
Density 0.9100g/mL
Gallium (Ga) 0.000002% max.
Merck Index 15, 491
Copper (Cu) 0.00001% max.
Indium (In) 0.000002% max.
Iron (Fe) 0.00001% max.
Lead (Pb) 0.000005% max.
Lithium (Li) 0.000002% max.
Formula Weight 35.05
Magnesium (Mg) 0.00005% max.
Manganese (Mn) 0.00001% max.
Grade Analysis
Molybdenum (Mo) 0.00001% max.
Nickel (Ni) 0.000005% max.
Packaging Glass bottle
Phosphate 0.00005% max.
Non_volatile Matter 0.002% max.
Potassium (K) 0.0001% max.
Platinum (Pt) 0.00001% max.
Reducing Substances 0.0005% max.
Sodium (Na) 0.0001% max.
Solubility Solubility in water: soluble.
Specific Gravity 0.91
Strontium (Sr) 0.0001% max.
Sulfate 0.0002% max.
Sulfide 0.0002% max.
Thallium (Tl) 0.000005% max.
Titanium (Ti) 0.00001% max.
Tin (Sn) 0.00001% max.
Zinc (Zn) 0.00001% max.
Melting Point -57.0°C
Boiling Point 38.0°C
Chemical Name or Material Ammonium hydroxide
Show More Show Less

GHS P Statement
May cause respiratory irritation.
Very toxic to aquatic life.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger