Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
25L
Packaging:
Glass bottle
Plastic drum
1336-21-6, 7732-18-5
H5NO
35.05
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
14923
CHEBI:18219
azanium;hydroxide
[NH4+].[OH-]
1336-21-6, 7732-18-5
H5NO
35.05
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
14923
CHEBI:18219
azanium;hydroxide
[NH4+].[OH-]
Linear Formula NH4OH
Density 0.9100g/mL
Formula Weight 35.05
Grade Analysis
Packaging Glass bottle
Solubility Solubility in water: soluble.
Specific Gravity 0.91
Melting Point -57.0°C
Boiling Point 38.0°C
Quantity 1L
Chemical Name or Material Ammonium hydroxide
Show More Show Less

GHS P Statement
May cause respiratory irritation.
Very toxic to aquatic life.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger