Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Hydroxide, Reagent, ACS, 28-30%, Spectrum™ Chemical
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
6 x 500 mL
13.5 L
20 L
4 x 2.5 L
4 x 4 L
6 x 2.5 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum
Poly-Coated Glass Bottle
Description
Spectrum™ Chemical Ammonium Hydroxide, Reagent, ACS is a solution of amonia in water. Aqueous amonia, as it′s otherwise known is used as a complexant and base in qualitative inorganic analysis. It becomes a deep blue color with copper solutions. Also used to dissolve silver oxide residues, such as the ones formed from Tollens′ reagent. It is also commmonly used in numerous household cleaning agents, such as window cleaning formula. As an ACS grade Reagent, Spectrum Chemical manufactured Ammonium Hydroxide is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity.
Specifications
CAS Max % | 100% |
UN Number | UN2672 |
Quantity | 20 L |
Percent Purity | 28 to 30% |
Grade | ACS |
Packaging | Poly Drum |
Residue after Ignition | 0.002% |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction