Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, Reagent, ACS, 28-30%, Spectrum™

Click to view available options
Quantity:
100 mL
13.5 L
2.5 L
20 L
200 L
4 L
4 x 2.5 L
4 x 4 L
500 mL
6 x 2.5 L
6 x 500 mL
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum
Poly-Coated Glass Bottle
1336-21-6
H5NO
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide
[NH4+].[OH-]
1336-21-6
H5NO
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide
[NH4+].[OH-]

A1195, 1336-21-6, NH4OH

Spectrum™ Ammonium Hydroxide, Reagent, ACS is a solution of amonia in water. Aqueous amonia, as it′s otherwise known is used as a complexant and base in qualitative inorganic analysis. It becomes a deep blue color with copper solutions. Also used to dissolve silver oxide residues, such as the ones formed from Tollens′ reagent. It is also commmonly used in numerous household cleaning agents, such as window cleaning formula. As an ACS grade Reagent, Spectrum Chemical manufactured Ammonium Hydroxide is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity.
Quantity 13.5 L
CAS Max % 100%
Chloride 0.5 ppm
UN Number UN2672
Heavy Metals (as Pb) 0.5 ppm
Percent Purity 28 to 30%
Packaging Poly Drum
Phosphate 2 ppm
Nitrate 2 ppm
Residue after Ignition 0.002%
Sulfate 2 ppm
Show More Show Less