Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium iodate, for analysis, ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Plastic bottle
13446-09-8
H4INO3
192.94
MFCD00049684
ZRDJERPXCFOFCP-UHFFFAOYSA-N
ammonium iodate,azanium iodate,iodic acid hio3 , ammonium salt,acmc-1bqbr,ammonium iodate 50g,iodic acid hio3 , ammonium salt 1:1
166805
azanium;iodate
[NH4+].[O-]I(=O)=O
13446-09-8
H4INO3
192.94
MFCD00049684
ZRDJERPXCFOFCP-UHFFFAOYSA-N
ammonium iodate,azanium iodate,iodic acid hio3 , ammonium salt,acmc-1bqbr,ammonium iodate 50g,iodic acid hio3 , ammonium salt 1:1
166805
azanium;iodate
[NH4+].[O-]I(=O)=O
Oxidation reagent
Solubility Information Solubility in water: 2g/L (15°C)
Formula Weight 192.94
Physical Form Powder
Percent Purity ≥99.0%
Grade Analysis
Heavy Metals (as Pb) 5ppm max.
Insoluble in Water 0.005% max.
Insoluble Matter 0.005% max. (in water)
Iodide 0.001% max.
Iron (Fe) 0.001% max.
Packaging Plastic bottle
Sulfate 0.005% max
Color White
Quantity 50g
Chemical Name or Material Ammonium iodate
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May intensify fire; oxidizer.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 236-592-4