Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium iodide, 99+%, for analysis, Acros Organics

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
12027-06-4
H4IN
144.94
MFCD00011424
UKFWSNCTAHXBQN-UHFFFAOYSA-N
ammonium iodide,azanium iodide,unii-oz8f027ldh,ammonium iodide nh4 i,oz8f027ldh,ammonium-iodid,ammonium jodatum,ammoniumiodid,hsdb 476,nh4i
25487
CHEBI:63040
azanium;iodide
[NH4+].[I-]
12027-06-4
H4IN
144.94
MFCD00011424
UKFWSNCTAHXBQN-UHFFFAOYSA-N
ammonium iodide,azanium iodide,unii-oz8f027ldh,ammonium iodide nh4 i,oz8f027ldh,ammonium-iodid,ammonium jodatum,ammoniumiodid,hsdb 476,nh4i
25487
CHEBI:63040
azanium;iodide
[NH4+].[I-]
Additional Information this product may turn yellow on storage
Assay Percent Range 99% min. (Argentometry)
Linear Formula NH4I
Merck Index 15, 526
Ash 0.05% max.
Solubility Information Solubility in water: 1770g/L (25°C). Other solubilities: 2500g/L (100°C),270g/L alcohol- 667g/L glycerol,400g/L methanol
Barium (Ba) 20ppm max.
Formula Weight 144.94
Physical Form Crystalline Powder
Percent Purity 99+%
Grade Analysis
Calcium (Ca) 20ppm max.
Chloride and Bromide 0.05% max. (as Cl)
Density 2.5100g/mL
Heavy Metals (as Pb) 10ppm max.
Iron (Fe) 5ppm max.
Packaging Glass bottle
Phosphate 10ppm max.
Solubility Solubility in water: 1770g/L (25°C). Other solubilities: 2500g/L (100°C), 270g/L alcohol - 667g/L glycerol, 400g/L methanol
Specific Gravity 2.51
Sulfate 100ppm max.
Color White to Yellow
Melting Point 551.0°C
pH 4.5 to 6.5 (5% soln. at 20°C)
Quantity 100g
Chemical Name or Material Ammonium iodide, p.a.
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

GHS Signal Word: Warning

EINECSNumber : 234-717-7

TSCA : TSCA