Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium iodide, ACS reagent, Acros Organics

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Glass bottle
12027-06-4
H4IN
144.94
UKFWSNCTAHXBQN-UHFFFAOYSA-N
ammonium iodide,azanium iodide,unii-oz8f027ldh,ammonium iodide nh4 i,oz8f027ldh,ammonium-iodid,ammonium jodatum,ammoniumiodid,hsdb 476,nh4i
25487
CHEBI:63040
azanium;iodide
[NH4+].[I-]
12027-06-4
H4IN
144.94
UKFWSNCTAHXBQN-UHFFFAOYSA-N
ammonium iodide,azanium iodide,unii-oz8f027ldh,ammonium iodide nh4 i,oz8f027ldh,ammonium-iodid,ammonium jodatum,ammoniumiodid,hsdb 476,nh4i
25487
CHEBI:63040
azanium;iodide
[NH4+].[I-]
Linear Formula NH4I
Merck Index 15,526
Solubility Information Solubility in water: 1770g/L (25°C). Other solubilities: 2500g/L (100°C),270g/L alcohol- 667g/L glycerol,400g/L methanol
Formula Weight 144.94
Grade ACS Reagent
Density 2.5100g/mL
Insoluble Matter 0.005% max
Packaging Glass bottle
Phosphate 0.001% max
Residue after Ignition 0.05% max
Solubility Solubility in water: 1770g/L (25°C). Other solubilities: 2500g/L (100°C), 270g/L alcohol - 667g/L glycerol, 400g/L methanol
Specific Gravity 2.51
Sulfate 0.05% max
Melting Point 551.0°C
Quantity 50g
Chemical Name or Material Ammonium iodide
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

GHS Signal Word: Warning

EINECSNumber : 234-717-7