Ammonium iron(II) sulfate, Dry, Acculute Standard Volumetric Solution, 0.1N, Alfa Aesar™

Air and Light Sensitive

Manufacturer: Alfa Aesar 88584LS

Quantity 6 units

 View more versions of this product

Catalog No. AA88584LS

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7783-85-9
    Physical Form Crystals
    Assay Percent Range 0.1N
    Molecular Formula H8FeN2O8S2
    Formula Weight 284.04
    MDL Number MFCD00051039
    Synonym Ammonium iron(II) sulfate, hexahydrate
    Chemical Name or Material Ammonium iron(II) sulfate

    Specifications

    Physical Form Crystals
    Formula Weight 284.04
    Quantity 6 units
    Color Pale blue-green
    EINECS Number 233-382-4
    Odor Odorless
    Assay Percent Range 0.1N
    Molecular Formula H8FeN2O8S2
    MDL Number MFCD00051039
    CAS 7783-85-9
    Synonym Ammonium iron(II) sulfate, hexahydrate
    Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Ammonium iron(II) sulfate

    H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit