Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium iron(III) hexacyanoferrate(II) hydrate, tech., Thermo Scientific Chemicals

Catalog Number p-7026103
Click to view available options
Quantity:
25 g
100 g
25869-00-5
C6H4Fe2N7
285.837
MFCD01726733
ILZWGESBVHGTRX-UHFFFAOYSA-O
ammonium iron iii hexacyanoferrate ii, iron 3+ ammonium ferrocyanide, azanium;iron 2+ ;iron 3+ ;hexacyanide, ammonium iron iii hexacyanoferrate ii hydrate, ammonium iron iii hexacyanoferrate ii hydrate, tech., ammonium iron iii hexacyanoferrate ii giese salt
23620040
azanium;iron(2+);iron(3+);hexacyanide
[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[NH4+].[Fe+2].[Fe+3]
25869-00-5
C6H4Fe2N7
285.837
MFCD01726733
ILZWGESBVHGTRX-UHFFFAOYSA-O
ammonium iron iii hexacyanoferrate ii, iron 3+ ammonium ferrocyanide, azanium;iron 2+ ;iron 3+ ;hexacyanide, ammonium iron iii hexacyanoferrate ii hydrate, ammonium iron iii hexacyanoferrate ii hydrate, tech., ammonium iron iii hexacyanoferrate ii giese salt
23620040
azanium;iron(2+);iron(3+);hexacyanide
[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[NH4+].[Fe+2].[Fe+3]

It is employed as a intermediate for pharmaceuticals. Also used in decorative cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceuticals. Also used in decorative cosmetics.

Solubility
Soluble in water and mineral acids. Insoluble in alkali.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 25 g
Merck Index 14,7910
Solubility Information Soluble in water and mineral acids. Insoluble in alkali.
Formula Weight 285.83 (Anhydrous)
Grade Technical
Chemical Name or Material Ammonium iron(III) hexacyanoferrate(II) hydrate
EINECSNumber 247-304-1
RTECSNumber LJ8140000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit