Learn More
Alfa Aesar™ Ammonium iron(III) oxalate hydrate
In photography, blueprints
Manufacturer:  Alfa Aesar™ 4211230
Quantity 250g
 View more versions of this product
Catalog No. AA4211230
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
13268-42-3
MFCD00013071
14,517
Ammonium iron(III) oxalate hydrate
374.02 (Anhydrous)
UN3288
Ferric ammonium oxalate; Iron(III) ammonium oxalate
Specifications
13268-42-3
374.02 (Anhydrous)
UN3288
14,517
Ferric ammonium oxalate; Iron(III) ammonium oxalate
Ammonium iron(III) oxalate hydrate
Powder
MFCD00013071
250g
Light Sensitive & Hygroscopic
Very soluble in water; Insoluble in alcohol
Safety and Handling
SDS & Safety Documents
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
TSCA : TSCA