Alfa Aesar™ Ammonium iron(III) oxalate hydrate

In photography, blueprints

Manufacturer: Alfa Aesar™ 4211218

Quantity 50g

 View more versions of this product

Catalog No. AA4211218

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13268-42-3
    Formula Weight 374.02 (Anhydrous)
    MDL Number MFCD00013071
    UN Number UN3288
    Merck Index 14,517
    Synonym Ferric ammonium oxalate; Iron(III) ammonium oxalate
    Chemical Name or Material Ammonium iron(III) oxalate hydrate

    Specifications

    Formula Weight 374.02 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P280-P301+P312-P312-P363-P322-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific measures (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00013071
    UN Number UN3288
    Merck Index 14,517
    Sensitivity Light Sensitive & Hygroscopic
    Synonym Ferric ammonium oxalate; Iron(III) ammonium oxalate
    Solubility Information Very soluble in water; Insoluble in alcohol
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    CAS 13268-42-3
    Quantity 50g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312
    Harmful if swallowed.
    Harmful in contact with skin.
    Chemical Name or Material Ammonium iron(III) oxalate hydrate

    GHS H Statement
    H302-H312
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P280-P301+P312-P312-P363-P322-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific measures (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit