Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Iron(Iii) Sulfate Dodecahydrate, 99%, ACS Reagent, Honeywell™ Fluka™

Click to view available options
Quantity:
25g
500g
2.5kg
Packaging:
Poly Bottle
7783-83-7
FeH28NO20S2
482.176
MFCD00150004
LCPUDZUWZDSKMX-UHFFFAOYSA-K
ammonium ferric sulfate,ferric ammonium sulfate dodecahydrate,iron ammonium sulfate dodecahydrate,sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate,ferric ammonium sulfate, acs,azanium iron 3+ disulfate dodecahydrate,ferric iron ammonium dodecahydrate disulfate,azanium iron +3 cation disulfate dodecahydrate,ammonium iron iii sulfate dodecahydrate, acs 100g
197097
azanium;iron(3+);disulfate;dodecahydrate
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
7783-83-7
FeH28NO20S2
482.176
MFCD00150004
LCPUDZUWZDSKMX-UHFFFAOYSA-K
ammonium ferric sulfate,ferric ammonium sulfate dodecahydrate,iron ammonium sulfate dodecahydrate,sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate,ferric ammonium sulfate, acs,azanium iron 3+ disulfate dodecahydrate,ferric iron ammonium dodecahydrate disulfate,azanium iron +3 cation disulfate dodecahydrate,ammonium iron iii sulfate dodecahydrate, acs 100g
197097
azanium;iron(3+);disulfate;dodecahydrate
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]

ACS Reagent, 99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Linear Formula NH4Fe(SO4)2 · 12H2O
UN Number NONH for all modes of transport
Formula Weight 482.19g/mol
Percent Purity 99%
Grade ACS Reagent
Packaging Poly Bottle
Quantity 2.5kg
Chemical Name or Material Ammonium Iron(Iii) Sulfate Dodecahydrate

EINECSNumber : 233-382-4

RTECSNumber : WS5900000