Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium iron(III) sulfate dodecahydrate, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
50g
100g
500g
2.5kg
Packaging:
Plastic bottle
7783-83-7
H4FeNO8S2·12H2O
482.19
MFCD00150004
LCPUDZUWZDSKMX-UHFFFAOYSA-K
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
197097
azanium;iron(3+);disulfate;dodecahydrate
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
7783-83-7
H4FeNO8S2·12H2O
482.19
MFCD00150004
LCPUDZUWZDSKMX-UHFFFAOYSA-K
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
197097
azanium;iron(3+);disulfate;dodecahydrate
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
Assay Percent Range 99% min. (Iodometry)
Linear Formula (NH4)Fe(SO4)2·12H2O
Merck Index 15, 515
Solubility Information Solubility in water: 1240g/L water (25°C). Other solubilities: practically insoluble in alcohol
Formula Weight 482.19
Physical Form Adhering Crystals or Powder
Percent Purity 99+%
Grade Analysis
Chloride 5ppm max
Chlorine (Cl) 5ppm max.
Copper (Cu) 10ppm max.
Density 1.7100g/mL
Iron (Fe2) 10ppm max.
Nitrate 100ppm max.
Packaging Plastic bottle
Phosphate 20ppm max.
Solubility Solubility in water: 1240g/L water (25°C). Other solubilities: practically insoluble in alcohol
Specific Gravity 1.71
Trace Analysis 10ppm max.
Boiling Point 230.0°C
Color Gray-Purple
Melting Point 39.0°C to 41.0°C
Quantity 2.5kg
Chemical Name or Material Ammonium iron(III) sulfate dodecahydrate, p.a.
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec

GHS Signal Word: Warning

RTECSNumber : WS5900000