Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium meta-Vanadate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

7803-55-6
H4NO3V
116.977
UNTBPXHCXVWYOI-UHFFFAOYSA-O
516859

High quality chemicals for laboratory and specialized industrial use

SMILES [NH4+].[O-][V](=O)=O
IUPAC Name azanium;oxido(dioxo)vanadium
Formula Weight 116.98
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.33kg
Quantity 500g
Chemical Name or Material Ammonium meta-Vanadate
Grade Baker Analyzed™ ACS Reagent