Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium meta-Vanadate (Certified ACS), Fisher Chemical

Catalog Number a713500
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
7803-55-6
H4NO3V
116.98
MFCD00011430
YBVKNHXQSRDWAA-UHFFFAOYSA-M
ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
516859
vanadiumoylol amine
N.O[V](=O)=O
7803-55-6
H4NO3V
116.98
MFCD00011430
YBVKNHXQSRDWAA-UHFFFAOYSA-M
ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
516859
vanadiumoylol amine
N.O[V](=O)=O
Melting Point 200°C
Color Green
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥99.0 %
Formula Weight 116.98
Percent Purity ≥99.0%
Grade Certified ACS
Identification Pass Test
Packaging Glass Bottle
Chemical Name or Material Ammonium meta-Vanadate
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
May be fatal if inhaled. Irritating to eyes, respiratory system and skin. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:4
Flammability:1
Instability:1
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit