Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Metatungstate, Hydrate, Spectrum™

Click to view available options
Quantity:
25 g
100 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
12028-48-7
12028-48-7
Grade Reagent
CAS Max % 1
Packaging Poly Pail
Tungsten (W) 0.713
Quantity 5 kg

CAUTION: For use by qualified personnel for research and development only. Read and understand the label and Safety Data Sheet (SDS) prior to use.