Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium metavanadate, ACS reagent, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
7803-55-6
H4NO3V
116.98
UNTBPXHCXVWYOI-UHFFFAOYSA-O
ammonium metavanadate,ammonium vanadate v,ammonium monovanadate,unii-fl85px638g,vanadate vo31-, ammonium,vanadate vo31-, ammonium 1:1,ammonium vanadium oxide,ammoniummetavanadate,vanadic acid, hvo3 , ammonium salt,ammonium meta-vanadate
516859
azanium;oxido(dioxo)vanadium
[NH4+].[O-][V](=O)=O
7803-55-6
H4NO3V
116.98
UNTBPXHCXVWYOI-UHFFFAOYSA-O
ammonium metavanadate,ammonium vanadate v,ammonium monovanadate,unii-fl85px638g,vanadate vo31-, ammonium,vanadate vo31-, ammonium 1:1,ammonium vanadium oxide,ammoniummetavanadate,vanadic acid, hvo3 , ammonium salt,ammonium meta-vanadate
516859
azanium;oxido(dioxo)vanadium
[NH4+].[O-][V](=O)=O
Linear Formula NH4VO3
Merck Index 15,565
Solubility Information Solubility in water: 5.1g/L (20°C). Other solubilities: insoluble in ethanol,69.5g/L (96°C)
Formula Weight 116.98
Physical Form Crystalline Solid or Powder
Percent Purity ≥99.0% (Titrimetry other)
Grade ACS Reagent
Carbonate passes test
Chloride 0.2 % max.
Packaging Glass bottle
Solubility (in Ammonium hydroxide) Passes test
Sulfate 0.05 % max.
Color White to Yellow
Melting Point 200.0°C
Quantity 100g
Chemical Name or Material Ammonium metavanadate
Show More Show Less

GHS H Statement
Suspected of damaging the unborn child.
Causes damage to organs through prolonged or repeated exposure if inhaled.
Toxic if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Toxic to aquati

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Danger

EINECSNumber : 232-261-3