Learn More
Ammonium Molybdate, 15% w/v, Reagents
Ammonium Molybdate, 15% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CA0058001A
| Quantity | 1 L |
|---|
Chemical Identifiers
| 7732-18-5 , 12054-85-2 | |
| 18.02 | |
| water |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| O |
Specifications
| Colorless to Light Green | |
| 100°C | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 15% | |
| Poly Bottle | |
| 5 to 6 | |
| Non-sterile | |
| Ammonium Molybdate, 15% w/v |
| 7732-18-5 , 12054-85-2 | |
| H2O | |
| O | |
| 18.02 | |
| Reagent | |
| 1.0902 | |
| 1 L | |
| Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.