Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium molybdate, 99.998%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
100g
Packaging:
Glass bottle
13106-76-8
H8MoN2O4
MFCD00011596
13106-76-8
H8MoN2O4
MFCD00011596
For ultra trace analysis, Oxidation reagent
Assay Percent Range 99.998%
Linear Formula (NH4)2MoO4
Formula Weight 196.01
Physical Form Powder
Grade Trace Metal Basis
Packaging Glass bottle
Total Trace Metal Impurities 20ppm max.
Color White
Quantity 100g
Chemical Name or Material Ammonium molybdate

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.