Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Ammonium molybdate (di), 4% w/v aq. soln.
Click to view available options
Quantity:
500mL
1L
Specifications
Concentration | 4% w/v aq. soln. |
Formula Weight | 339.95 |
Physical Form | Liquid |
Quantity | 500mL |
Chemical Name or Material | Ammonium molybdate (di) |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures