Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Molybdate, Tetrahydrate, USP, 99.3-101.8%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
12054-85-2
H6MoNO2
148.00
XCOMAVCHADVANQ-UHFFFAOYSA-O
ammonium hydrate molybdenum oxidandiide
[NH4+].O.[O--].[Mo]
12054-85-2
H6MoNO2
148.00
XCOMAVCHADVANQ-UHFFFAOYSA-O
ammonium hydrate molybdenum oxidandiide
[NH4+].O.[O--].[Mo]

A1675, 12054-85-2, (NH4)6Mo7O24•4H2O

Spectrum™ Ammonium Molybdate, Tetrahydrate, USP is used therapeutically in the treatment of copper toxicosis. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Percent Purity 99.3 to 101.8%
Chloride 20 ppm
Insoluble Matter 0.005%
Packaging Poly Pail
Phosphate 5 ppm
Sulfate 200 ppm
Quantity 2.5 kg