Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Nitrate, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5383200
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
6484-52-2
H4N2O3
80.04
MFCD00011425
DVARTQFDIMZBAA-UHFFFAOYSA-O
ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
22985
CHEBI:63038
ammonium nitrate
[NH4+].[O-][N+]([O-])=O
This item is not returnable. View return policy
6484-52-2
H4N2O3
80.04
MFCD00011425
DVARTQFDIMZBAA-UHFFFAOYSA-O
ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
22985
CHEBI:63038
ammonium nitrate
[NH4+].[O-][N+]([O-])=O

High quality chemicals for laboratory and specialized industrial use

Quantity 500 g
Formula Weight 80.04
Grade Meets ACS reagent chemical requirements
Packaging Poly Bottle
Specific Gravity 1L = 1.73kg
Chemical Name or Material Ammonium Nitrate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit