Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Nitrate, Granular, Reagent, ACS, 95%, Spectrum™

Click to view available options
Quantity:
25 g
500 g
2.5 kg
6 x 500 g
12 kg
25 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6484-52-2
H4N2O3
80.04
DVARTQFDIMZBAA-UHFFFAOYSA-O
ammonium nitrate
[NH4+].[O-][N+]([O-])=O
6484-52-2
H4N2O3
80.04
DVARTQFDIMZBAA-UHFFFAOYSA-O
ammonium nitrate
[NH4+].[O-][N+]([O-])=O

A1215, 6484-52-2, NH4NO3

Spectrum™ Ammonium Nitrate, Granular, Reagent, ACS is a white crystalline solid which is highly soluble in water. It is most commonly used in agriculture as a nitrogen fertilizer. It is a main component in some explosives and also finds use in instant cold packs as hydrating the salt is an endothermic process. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
CAS Max % 1
UN Number UN1942
Percent Purity 95%
Chloride 5 ppm
Heavy Metals (as Pb) 5 ppm
Insoluble Matter 5e-005
Packaging Poly Pail
Phosphate 5 ppm
pH Range 4.5-6.0
Residue after Ignition 0.0001
Sulfate 2e-005
Quantity 12 kg
Show More Show Less