Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Nitrate, Purified, Prill, Reagents

Catalog No. p-200021201
Click to view available options
Quantity:
500 g
3 kg
12 kg
25 kg

Ammonium Nitrate, Purified, Prill, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6484-52-2
Molecular Formula H4N2O3
Molecular Weight (g/mol) 80.04
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
Synonym Nitric Acid ammonium salt
IUPAC Name ammonium nitrate
SMILES [NH4+].[O-][N+]([O-])=O

Specifications

Boiling Point 210°C
Melting Point 170°C
Color Colorless/White
Physical Form Solid
Quantity 12 kg
Grade Pure
Packaging Poly Pail
Specific Gravity 1.72
Chemical Name or Material Ammonium Nitrate, Purified, Prill
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.