Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium oxalate monohydrate, 98%, Thermo Scientific Chemicals

Catalog Number p-7080647
Click to view available options
Quantity:
100 g
500 g
2500 g
6009-70-7
C2H10N2O5
142.111
MFCD00149694
MSMNVXKYCPHLLN-UHFFFAOYSA-N
ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
516808
diazanium;oxalate;hydrate
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O
6009-70-7
C2H10N2O5
142.111
MFCD00149694
MSMNVXKYCPHLLN-UHFFFAOYSA-N
ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
516808
diazanium;oxalate;hydrate
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O

Ammonium oxalate monohydrate is widely utilized as a buffering reagent and a dispersant to determine the surface interaction of platelets. It finds an application to study its acute poisoning effect on sheep and to investigate the formation of oxalate monoalkylamide in the human lens. It is also used in the detection and determination of calcium, lead, fluoride and rare earth metals. It is employed as chelators and forms complexes with metals. It acts as reducing agent in gold extraction and is an active ingredient of ferrous metal surface polishing fluid. It is a promoting agent in production of cobalt oxide and nickel oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium oxalate monohydrate is widely utilized as a buffering reagent and a dispersant to determine the surface interaction of platelets. It finds an application to study its acute poisoning effect on sheep and to investigate the formation of oxalate monoalkylamide in the human lens. It is also used in the detection and determination of calcium, lead, fluoride and rare earth metals. It is employed as chelators and forms complexes with metals. It acts as reducing agent in gold extraction and is an active ingredient of ferrous metal surface polishing fluid. It is a promoting agent in production of cobalt oxide and nickel oxide.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY
Melting Point 70°C (decomposition)
Density 1.5 g/mL
Assay Percent Range >99%
Linear Formula (COONH4)2·H2O
Quantity 100 g
UN Number UN3288
Beilstein 3759830
Merck Index 14,537
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 142.11 (124.10Anhydrous)
Percent Purity 98%
Chemical Name or Material Ammonium oxalate monohydrate
Show More Show Less
Hazard Category H302+H312-H320-H335-H372
Hazard Statement GHS H Statement
H302-H312
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 238-135-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit