Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium oxalate monohydrate, 99.5%, for analysis, ACROS Organics™

Click to view available options
Quantity:
500g
1kg
Packaging:
Plastic bottle
6009-70-7
C2H8N2O4·H2O
142.11
MFCD00149694
MSMNVXKYCPHLLN-UHFFFAOYSA-N
ammonium oxalate monohydrate,ammonium oxalate monohydrate, acs,ethanedioic acid, diammonium salt, monohydrate,diazanium oxalate hydrate,acmc-1aw90,ksc493e5j,diammonium hydrate oxalate,oxalic acid ammonium salt monohydrate
516808
diazanium;oxalate;hydrate
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O
6009-70-7
C2H8N2O4·H2O
142.11
MFCD00149694
MSMNVXKYCPHLLN-UHFFFAOYSA-N
ammonium oxalate monohydrate,ammonium oxalate monohydrate, acs,ethanedioic acid, diammonium salt, monohydrate,diazanium oxalate hydrate,acmc-1aw90,ksc493e5j,diammonium hydrate oxalate,oxalic acid ammonium salt monohydrate
516808
diazanium;oxalate;hydrate
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O
Assay Percent Range 99.50%
Linear Formula (CO2NH4)2·H2O
Merck Index 15, 534
Solubility Information Solubility in water: 45g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 142.11
Physical Form Crystalline Powder
Percent Purity 99.5%
Grade Analysis
Chloride 20ppm max.
Heavy Metals (as Pb) 10ppm max.
Iron (Fe) 2ppm max.
Packaging Plastic bottle
Solubility Solubility in water: 45g/L (20°C). Other solubilities: slightly soluble in alcohol
Sulfate 50ppm max.
Color White
Melting Point 70.0°C
Quantity 1kg
Chemical Name or Material Ammonium oxalate monohydrate, p.a.
Show More Show Less

GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning