Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium oxalate monohydrate, 99.5%, for analysis, ACROS Organics™
Click to view available options
Quantity:
500g
1kg
Packaging:
Plastic bottle
Specifications
Assay Percent Range | 99.50% |
Linear Formula | (CO2NH4)2·H2O |
Merck Index | 15, 534 |
Solubility Information | Solubility in water: 45g/L (20°C). Other solubilities: slightly soluble in alcohol |
Formula Weight | 142.11 |
Physical Form | Crystalline Powder |
Percent Purity | 99.5% |
Grade | Analysis |
Chloride | 20ppm max. |
Heavy Metals (as Pb) | 10ppm max. |
Show More |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning