Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Oxalate Monohydrate, ACS Reagent, Reagents

Catalog No. p-200021190
Click to view available options
Quantity:
100 g
500 g
3 kg

Ammonium Oxalate Monohydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6009-70-7
Molecular Formula C2H10N2O5
Molecular Weight (g/mol) 142.11
InChI Key MSMNVXKYCPHLLN-UHFFFAOYSA-N
Synonym Diammonium ethanedioic Acid monohydrate
IUPAC Name oxalic acid diamine hydrate
SMILES N.N.O.OC(=O)C(O)=O

Specifications

Boiling Point 365°C
Melting Point 70°C
Color Colorless/White
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.5
Chemical Name or Material Ammonium Oxalate Monohydrate, ACS Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.