Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Oxalate, Saturated Solution, 5%, Spectrum™

Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
Grade Chemical Solution
CAS 6009-70-7,7732-18-5
Percent Purity 0.05
Packaging Poly Bottle
Quantity 1 L

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.