Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Perchlorate, Spectrum™

Click to view available options
Quantity:
25 g
500 g
2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
7790-98-9
7790-98-9
Grade Reagent
CAS Max % 1
Chloride and Chlorate 3e-05
Heavy Metals 5e-06
Insoluble Matter 5e-05
Iron (Fe) 5e-06
Packaging Poly Bottle
Sulfate 1e-05
pH 4.0 to 6.0
Quantity 2.5 kg

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.