Ammonium peroxydisulfate, ACS, 98.0% min, Alfa Aesar™

Manufacturer: Alfa Aesar 54106D9

Quantity 5 x 1kg

 View more versions of this product

Catalog No. AA54106D9

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7727-54-0
    Physical Form Lumps
    Assay Percent Range 98.0% min
    Molecular Formula H8N2O8S2
    Formula Weight 228.19
    MDL Number MFCD00003390
    Synonym Ammonium persulfate
    Chemical Name or Material Ammonium peroxydisulfate
    Grade ACS Grade

    Specifications

    Formula Weight 228.19
    Quantity 5 x 1kg
    EINECS Number 231-786-5
    Physical Form Lumps
    Assay Percent Range 98.0% min
    Molecular Formula H8N2O8S2
    MDL Number MFCD00003390
    CAS 7727-54-0
    Synonym Ammonium persulfate
    Chemical Name or Material Ammonium peroxydisulfate
    Grade ACS Grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit