Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Peroxydisulfate, Crystal, BiotechGrade, 98%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
7727-54-0
H8N2O8S2
228.19
ROOXNKNUYICQNP-UHFFFAOYSA-N
diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
7727-54-0
H8N2O8S2
228.19
ROOXNKNUYICQNP-UHFFFAOYSA-N
diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

A1227, 7727-54-0, (NH4)2S2O8

Spectrum™ Ammonium Peroxydisulfate, Crystal, BiotechGrade this highly water soluble oxidizing agent is a catalyst for acrylamide gel polymerization when used in conjunction with Tetramethylethylenediamine. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.
CAS Max % 100%
UN Number UN1444
Appearance Colorless to white
Percent Purity 98%
DNase RNase Protease act None detected
Heavy Metals (as Pb) 0.001%
Insoluble Matter 0.005%
Manganese (Mn) 0.5 ppm
Packaging Poly Bottle
Residue after Ignition 0.05%
Quantity 500 g
Show More Show Less