Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Persulfate, ≥98.0%, ACS Reagent, Honeywell™ Fluka™

Click to view available options
Quantity:
100 g
2.5 kg
4 x 2.5 kg
Packaging:
Poly Bottle
7727-54-0
H8N2O8S2
228.19
MFCD00003390
ROOXNKNUYICQNP-UHFFFAOYSA-N
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
62648
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
7727-54-0
H8N2O8S2
228.19
MFCD00003390
ROOXNKNUYICQNP-UHFFFAOYSA-N
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
62648
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

ACS Reagent,≥98.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Melting Point 120°C
Color Colorless
Physical Form Solid
Quantity 4 x 2.5 kg
Linear Formula (NH4)2S2O8
UN Number UN1444
IUPAC Name diazanium;sulfonatooxy sulfate
Formula Weight 228.2g/mol
Percent Purity ≥98.0%
Grade ACS Reagent
Packaging Poly Bottle
Residue after Ignition ≤0.05%
Density 1.98g/cm3; Vapor Density: 7.9 (vs air)
Chemical Name or Material Ammonium Persulfate
Show More Show Less

P220-P261-P280-P305 + P351 + P338-P342 + P311

H272-H302-H315-H317-H319-H334-H335

DOTInformation : 5.1/PGIII

EINECSNumber : 231-786-5

RTECSNumber : SE0350000