Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Persulfate, ACS Reagent (Ammonium Peroxydisulfate), Reagents
Click to view available options
Quantity:
100 mL
500 g
2.5 kg
12 kg
Specifications
Specifications
| Melting Point | 120°C to 160°C |
| Color | Colorless/White |
| Physical Form | Solid |
| Quantity | 2.5 kg |
| Grade | ACS |
| Packaging | Poly Bottle |
| Specific Gravity | 1.98 |
| Chemical Name or Material | Ammonium Persulfate, ACS Reagent (Ammonium Peroxydisulfate) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction