Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Persulfate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

7727-54-0
H8N2O8S2
228.19
ROOXNKNUYICQNP-UHFFFAOYSA-N
62648

High quality chemicals for laboratory and specialized industrial use

CAS 7727-54-0
Molecular Formula H8N2O8S2
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
IUPAC Name diazanium;sulfonatooxy sulfate
Molecular Weight (g/mol) 228.19
PubChem CID 62648
Formula Weight 228.2
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 1.98kg
Quantity 500g
Chemical Name or Material Ammonium Persulfate
Grade Baker Analyzed™ ACS Reagent
Show More Show Less