Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Persulfate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

7727-54-0
H8N2O8S2
228.19
ROOXNKNUYICQNP-UHFFFAOYSA-N
62648

High quality chemicals for laboratory and specialized industrial use

SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
IUPAC Name diazanium;sulfonatooxy sulfate
Formula Weight 228.2
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 1.98kg
Quantity 2.5kg
Chemical Name or Material Ammonium Persulfate
Grade Baker Analyzed™ ACS Reagent