Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Persulfate, PlusOne™, GE Healthcare

Click to view available options
Quantity:
25g
7727-54-0
H8N2O8S2
228.19
ROOXNKNUYICQNP-UHFFFAOYSA-N
ammonium persulfate,ammonium peroxydisulfate,diammonium peroxydisulfate,diammonium persulfate,ammoniumpersulfate,diammonium peroxodisulphate,diammonium peroxydisulphate,unii-22qf6l357f,ccris 1430,persulfate d'ammonium french
62648
diazanium;sulfonatooxy sulfate
[NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
7727-54-0
H8N2O8S2
228.19
ROOXNKNUYICQNP-UHFFFAOYSA-N
ammonium persulfate,ammonium peroxydisulfate,diammonium peroxydisulfate,diammonium persulfate,ammoniumpersulfate,diammonium peroxodisulphate,diammonium peroxydisulphate,unii-22qf6l357f,ccris 1430,persulfate d'ammonium french
62648
diazanium;sulfonatooxy sulfate
[NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]

Supports efficient polymerization of acrylamide at concentrations as low as 1mM

Assay Percent Range >98%
Formula Weight 228.2
Quantity 25g
Chemical Name or Material Ammonium Persulfate