Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium phosphate, dibasic, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
250g
500g
2.5kg
Packaging:
Plastic bottle
7783-28-0
H9N2O4P
132.06
MFCD00010891
MNNHAPBLZZVQHP-UHFFFAOYSA-N
diammonium phosphate,diammonium hydrogenphosphate,diammonium hydrogen phosphate,phosphoric acid, diammonium salt,fyrex,pelor,akoustan a,diammonium orthophosphate,diammonium acid phosphate,dibasic ammonium phosphate
24540
CHEBI:63051
diazanium;hydrogen phosphate
[NH4+].[NH4+].OP(=O)([O-])[O-]
7783-28-0
H9N2O4P
132.06
MFCD00010891
MNNHAPBLZZVQHP-UHFFFAOYSA-N
diammonium phosphate,diammonium hydrogenphosphate,diammonium hydrogen phosphate,phosphoric acid, diammonium salt,fyrex,pelor,akoustan a,diammonium orthophosphate,diammonium acid phosphate,dibasic ammonium phosphate
24540
CHEBI:63051
diazanium;hydrogen phosphate
[NH4+].[NH4+].OP(=O)([O-])[O-]
Assay Percent Range 99+%
Linear Formula (NH4)2HPO4
Merck Index 15, 540
Solubility Information Solubility in water: 58g/100mL (20°C). Other solubilities: practically insoluble in alcohol and acetone
Formula Weight 132.06
Physical Form Adhering Crystals
Percent Purity 99+%
Grade Analysis
Chloride 5ppm max.
Density 1.61
Heavy Metals (as Pb) 10ppm max.
Iron (Fe) 10ppm max.
Nitrate 100ppm max.
Packaging Plastic bottle
Potassium (K) 100ppm max.
Sodium (Na) 100ppm max.
Solubility Solubility in water: 58g/100mL (20°C). Other solubilities: practically insoluble in alcohol and acetone
Sulfate 200ppm max.
Color White
pH 7.9 to 8.3 (5% soln. at 20°C)
Quantity 500g
Chemical Name or Material Ammonium phosphate, dibasic, p.a.
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 231-987-8

TSCA : TSCA