Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium phosphate, dibasic, 99%, for biochemistry, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
Packaging:
Plastic bottle
7783-28-0
H9N2O4P
132.06
MNNHAPBLZZVQHP-UHFFFAOYSA-N
diammonium phosphate,diammonium hydrogenphosphate,diammonium hydrogen phosphate,phosphoric acid, diammonium salt,fyrex,pelor,akoustan a,diammonium orthophosphate,diammonium acid phosphate,dibasic ammonium phosphate
24540
CHEBI:63051
diazanium;hydrogen phosphate
[NH4+].[NH4+].OP(=O)([O-])[O-]
7783-28-0
H9N2O4P
132.06
MNNHAPBLZZVQHP-UHFFFAOYSA-N
diammonium phosphate,diammonium hydrogenphosphate,diammonium hydrogen phosphate,phosphoric acid, diammonium salt,fyrex,pelor,akoustan a,diammonium orthophosphate,diammonium acid phosphate,dibasic ammonium phosphate
24540
CHEBI:63051
diazanium;hydrogen phosphate
[NH4+].[NH4+].OP(=O)([O-])[O-]
Absorbance (c=1M, water),(c=1M, water) at 0nm,0.1 max. at 260nm,0.1 max. at 280nm
Assay Percent Range 99%
Linear Formula (NH4)2HPO4
Merck Index 15,54
Solubility Information Solubility in water: 58g/100mL (20°C). Other solubilities: practically insoluble in alcohol and acetone
Formula Weight 132.06
Percent Purity 99%
Grade Biochemistry
Packaging Plastic bottle
Solubility (1 M in water) Clear colorless
Quantity 1kg
Chemical Name or Material Ammonium phosphate, dibasic
Show More Show Less

Warning

EINECSNumber : 231-987-8