Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Phosphate Dibasic, Crystal, ACS, 98%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7783-28-0
7783-28-0
Grade ACS
CAS Max % 1
Percent Purity 0.98
Chloride 1e-05
Heavy Metals 1e-05
Insoluble Matter 5e-05
Iron (Fe) 1e-05
Nitrate 3e-05
Packaging Poly Bottle
Potassium (K) 5e-05
Sodium (Na) 5e-05
Sulfate 0.0001
pH 7.7 to 8.1
Quantity 500 g
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.