Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Phosphate, Dibasic, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

7783-28-0
H9N2O4P
132.056
MNNHAPBLZZVQHP-UHFFFAOYSA-N
24540
CHEBI:63051

High quality chemicals for laboratory and specialized industrial use

CAS 7783-28-0
Molecular Formula H9N2O4P
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
SMILES [NH4+].[NH4+].OP(=O)([O-])[O-]
IUPAC Name diazanium;hydrogen phosphate
Molecular Weight (g/mol) 132.056
PubChem CID 24540
ChEBI CHEBI:63051
Formula Weight 132.06
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 1.62kg
Quantity 2.5kg
Chemical Name or Material Ammonium Phosphate, Dibasic
Grade Baker Analyzed™ ACS Reagent
Show More Show Less