Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Phosphate Dibasic, FCC, 96-102%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7783-28-0
H9N2O4P
132.06
MNNHAPBLZZVQHP-UHFFFAOYSA-N
diammonium hydrogen phosphate
[NH4+].[NH4+].OP([O-])([O-])=O
7783-28-0
H9N2O4P
132.06
MNNHAPBLZZVQHP-UHFFFAOYSA-N
diammonium hydrogen phosphate
[NH4+].[NH4+].OP([O-])([O-])=O

AM184, 7783-28-0, (NH4)2HPO4

Spectrum™ Ammonium Phosphate Dibasic, FCC is a food ingredient used as an acidity regulator and raising agent. It is listed on the FDA′s Generally Recognized As Safe (GRAS) list of substances. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Arsenic (As) 3 mg/kg
Percent Purity 96 to 102%
Fluoride 10 mg/kg
Lead (Pb) 4 mg/kg
Packaging Poly Pail
Quantity 2.5 kg