Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Phosphate Dibasic, FCC, 96-102%, Spectrum™ Chemical
SDP

Catalog No. 18600643
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

AM184, 7783-28-0, (NH4)2HPO4

Spectrum™ Chemical Ammonium Phosphate Dibasic, FCC is a food ingredient used as an acidity regulator and raising agent. It is listed on the FDA′s Generally Recognized As Safe (GRAS) list of substances. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7783-28-0
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
IUPAC Name phosphoric acid diamine
SMILES N.N.OP(O)(O)=O

Specifications

CAS Max % 100%
Quantity 50 kg
Percent Purity 96 to 102%
Grade FCC
Packaging Fiber Drum
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.